Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate IL-15 Biotinylated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. BAM247
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM247 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM247 Supplier R&D Systems Supplier No. BAM247

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

IL-15 Monoclonal specifically detects IL-15 in Human∣Mouse∣Primate samples. It is validated for ELISA Detection∣ELISA Detection (Matched Antibody Pair)∣ELISA Development∣Luminex Development∣Western Blot.

Specifications

Antigen IL-15
Applications ELISA Detection (Matched Antibody Pair), ELISA Development, Luminex, Western Blot
Classification Monoclonal
Clone 34593
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. P40933
Gene Alias IL15, IL-15MGC9721, interleukin 15, interleukin-15
Gene Symbols IL15
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-15 Asn49-Ser162 Accession # P40933
Purification Method Protein A or G purified from ascites
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3600
Test Specificity Detects human IL-15 in ELISAs and Western blots. In ELISAs, this antibody shows no cross-reactivity or intereference with recombinant human (rh) IL-1 alpha , rmIL-1 alpha , rhIL-1 beta , rmIL-1 beta , rhIL-2, rhIL-2 R alpha , rhIL-2 R beta , rhIL-2 R gamma , rhIL-3, rmIL-3, rhIL-4, rmIL-4, rhIL-4 R, rhIL-5, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-7 R, rhIL-8, rhIL-9, rmIL-9, rmIL-13, and rhTGF-beta RII.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.