Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate VEGF Antibody, R&D Systems™
GSA_VAGREENER_CHOICESDP

Catalog No. MAB293100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB293100 100 μg
MAB293SP 25 μg
MAB293500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB293100 Supplier R&D Systems Supplier No. MAB293100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 53 publications

VEGF Monoclonal specifically detects VEGF in Bovine∣Complex Species Category∣Human∣Mouse∣Pig∣Primate∣Rat∣Rhesus Macaque samples. It is validated for Array Development∣Bioassay∣ELISA Capture (Matched Antibody Pair)∣ELISA Detection∣ELISA Development∣ELISA Microarray Development∣Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Neutralization∣Western Blot.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen VEGF
Applications Array, Bioassay, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Development, ELISA Microarray Development, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Neutralization, Western Blot
Classification Monoclonal
Clone 26503
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 10-60 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001165097.1
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF 165 Ala27-Arg191 Accession # NP_001165097.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity Detects human VEGF 165 and human VEGF 121 in ELISAs and Western blots. In ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) VEGF and recombinant rat (rr)VEGF and no cross-reactivity with recombinant human (rh) VEGF-D is obseved.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Bovine, Human, Mouse, Pig, Primate, Rat, Rhesus Macaque
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Supernatant
Isotype IgG2b
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.