Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate VEGF165 Biotinylated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. BAF293
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF293 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF293 Supplier R&D Systems Supplier No. BAF293

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

VEGF Polyclonal specifically detects VEGF in Human∣Primate samples. It is validated for Array Development∣Bioassay∣ELISA Detection∣ELISA Detection (Matched Antibody Pair)∣ELISA Development∣ELISA Development (Detection)∣ELISA Microarray Development∣Functional Assay∣Proximity Ligation Assay∣Quantum Dot∣TaqMan Protein Assay∣Western Blot.

Specifications

Antigen VEGF
Applications Array, Bioassay, ELISA Detection (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Development, ELISA Development (Detection), ELISA Microarray Development, Functional Assay, Proximity Ligation Assay, Quantum Dot, Protein Assay, Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. AAV38412
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF 165 (R&D Systems, Catalog # 293-VE) Ala27-Arg191 Accession # AAV38412
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity Detects human VEGF 165 in ELISAs and Western blots. In sandwich immunoassays, less than 15% cross-reactivity with recombinant human (rh) VEGF/PlGF is observed and less than 0.1% cross-reactivity with recombinant mouse (rm) PlGF-2, rhFlt-1, rhVEGF R2, rhPlGF, rhVEGF-C, rhVEGF-D, and rmVEGF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.