Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRKAA1 (NP_006242.4, 1 a.a.-550 a.a.)/PRKAB1 (NP_006244.2, 1 a.a. - 270 a.a.)/PRKAG1 (NP_002724.1, 1 a.a. - 331 a.a.) Full length Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Catalog No. 89958104 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-104 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-104 Supplier Abnova™ Supplier No. P5780
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit is one of the gamma regulatory subunits of AMPK. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Specifications

Accession Number NP_006242.4 (Gene ID : 5562); NP_006244.2 (Gene ID : 5564); NP_002724.1 (Gene ID : 5571)
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5562/5564/5571
Molecular Weight (g/mol) 90kDa
Name PRKAA1/PRKAB1/PRKAG1 (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AMPK/AMPKa1/MGC33776/MGC57364
Common Name PRKAA1
Gene Symbol PRKAA1
Biological Activity The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : SAMS peptide. ATP: 100 μM.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST-His
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 81% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.