Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRKACG (NM_002732) Full-length Recombinant Protein with GST expressed in Sf9 cells

Catalog No. 89958126 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-126 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-126 Supplier Abnova™ Supplier No. P4212

Used for Func, SDS-PAGE

Cyclic AMP-dependent protein kinase (PKA) consists of two catalytic subunits and a regulatory subunit dimer. This gene encodes the gamma form of its catalytic subunit. The gene is intronless and is thought to be a retrotransposon derived from the gene for the alpha form of the PKA catalytic subunit. [provided by RefSeq]

Specifications

Accession Number NM_002732
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5568
Molecular Weight (g/mol) 65kDa
Name PRKACG (Human) Recombinant Protein
Quality Control Testing 0.5 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KAPG/PKACg
Common Name PRKACG
Gene Symbol PRKACG
Biological Activity The specific activity of PKACgamma was determined to be 145nmol /min/mg as per activity assay protocol.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.