Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRKCH (P24723) Full-length Recombinant Protein with GST expressed in Sf9 cells

Catalog No. 89958181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-181 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-181 Supplier Abnova™ Supplier No. P4215

Used for Func, SDS-PAGE

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. It is a calcium-independent and phospholipids-dependent protein kinase. It is predominantly expressed in epithelial tissues and has been shown to reside specifically in the cell nucleus. This protein kinase can regulate keratinocyte differentiation by activating the MAP kinase MAPK13 (p38delta)-activated protein kinase cascade that targets CCAAT/enhancer-binding protein alpha (CEBPA). It is also found to mediate the transcription activation of the transglutaminase 1 (TGM1) gene. [provided by RefSeq]

Specifications

Accession Number P24723
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5583
Molecular Weight (g/mol) 103kDa
Name PRKCH (Human) Recombinant Protein
Quality Control Testing 1 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC26269/MGC5363/PKC-L/PKCL/PRKCL/nPKC-eta
Common Name PRKCH
Gene Symbol PRKCH
Biological Activity 86nmol phosphate incorporated into peptide substrate (ERMRPRKRQGSVRRRV) per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay).
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.