Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRKCI (NM_002740) Full-length Recombinant Protein expressed in Sf9 cells

Catalog No. 89958184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-184 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-184 Supplier Abnova™ Supplier No. P4189

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for Func, SDS-PAGE

This gene encodes a member of the protein kinase C (PKC) family of serine/threonine protein kinases. The PKC family comprises at least eight members, which are differentially expressed and are involved in a wide variety of cellular processes. This protein kinase is calcium-independent and phospholipid-dependent. It is not activated by phorbolesters or diacylglycerol. This kinase can be recruited to vesicle tubular clusters (VTCs) by direct interaction with the small GTPase RAB2, where this kinase phosphorylates glyceraldehyde-3-phosphate dehydrogenase (GAPD/GAPDH) and plays a role in microtubule dynamics in the early secretory pathway. This kinase is found to be necessary for BCL-ABL-mediated resistance to drug-induced apoptosis and therefore protects leukemia cells against drug-induced apoptosis. There is a single exon pseudogene mapped on chromosome X. [provided by RefSeq]

Specifications

Accession Number NM_002740
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5584
Molecular Weight (g/mol) 98kDa
Name PRKCI (Human) Recombinant Protein
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DXS1179E/MGC26534/PKCI/nPKC-iota
Common Name PRKCI
Gene Symbol PRKCI
Biological Activity 664nmol phosphate incorporated into CREBtide per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay in 25μL reaction volume).
Species Baculovirus
Recombinant Recombinant
Protein Tag None
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.