Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human PRMT1 Protein
SDP

Catalog No. NBC11844601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC11844601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC11844601 Supplier Novus Biologicals™ Supplier No. NBC1184460.1MG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-MBP tag and corresponding to the amino acids 1-353 of Human PRMT1 The Recombinant Human PRMT1 Protein is derived from E. coli. The Recombinant Human PRMT1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid in 40mM Tris pH 8.0, 100mM NaCl, 4mM MgCl2, 2mM DTT, 40% glycerol
Molecular Weight (g/mol) 84kDa
Purification Method Protein
Quantity 0.1 mg
Source Recombinant human PRMT1, fused to His-MBP tag at N-terminus, expressed in E. coli
Immunogen 1-353 aa, Human, His-MBP tagged, Recombinant, E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PRMT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.