Learn More
Specifications
Specifications
| Assay Range | 78.10 - 5000 pg/mL |
| Assay Sensitivity | 3.51 pg/mL |
| Product Line | R&D Systems |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Plasma (Heparin), Saliva, Serum, Tissue Lysates, Urine |
| Specificity | Natural and recombinant human Pro-Collagen I alpha 1. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1277 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | Alpha-1 type I collagen, COL1A1, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
