Learn More
R&D Systems™ Human Pro-Collagen II DuoSet ELISA
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Specifications
Specifications
| Description | Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, COL2A1, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC |
| Product Type | ELISA Kit |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Assay Sensitivity | 100 ng/mL |
| Cross Reactivity | Not observed |
| Format | 96-well Strip Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1280 |
| Sample Type | Cell Culture Supernatant, Serum, Plasma |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.