Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Pro-MMP-10 Quantikine ELISA Kit
SDP

Catalog No. DM1000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DM1000 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DM1000 Supplier R&D Systems™ Supplier No. DM1000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Pro-MMP-10 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Pro-MMP-10 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-10 in cell culture supernates (50 ul), serum (25 ul), heparin plasma (25 ul).

Specifications

Assay Range 78.10 to 5000 pg/mL
Assay Sensitivity 15.1 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum
Specificity Natural and Recombinant Human pro-MMP-10
For Use With (Application) ELISA
Gene ID (Entrez) 4319
Gene Symbol MMP10
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2
Target MMP-10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.