Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human ProANP Peptide, Synthetic, >95% Purity, 100 μg

Catalog No. 502710913
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0913 Supplier Abcam Supplier No. AB42339100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide with >95% purity for blocking applications.

  • Purity: >95%
  • Form: Lyophilized
  • Tested Applications: BL
  • Protein Type: Synthetic
  • Species Reactivity: Human

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Peptide-based Substrate
Target Molecule proANP (Human)
Form Lyophilized
Label or Dye Unconjugated
Shipping Condition Blue Ice
Content And Storage Store at -20°C.
Reactivity Human
Cross Reactivity Human
Quantity 100 μg
For Use With (Application) Blocking, Neutralization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.