Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human ProANP Peptide, Synthetic, >95% Purity, 100 μg

Catalog No. 502710922
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0922 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0922 Supplier Abcam Supplier No. AB42277100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide with >95% purity, suitable for blocking applications.

  • Purity: >95%
  • Form: Lyophilized
  • Storage buffer: 0.1% Acetic acid
  • Tested applications: BL
  • Species reactivity: Human
  • Protein type: Synthetic

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Blue Ice
Substrate Properties Peptide-based Substrate
Target Molecule proANP (Human)
Reactivity Human
Label or Dye Unconjugated
Buffer 0.1% acetic acid
Content And Storage Store at -20°C.
Form Lyophilized
Cross Reactivity Human
Formulation Lyophilized from 0.1% Acetic acid
Quantity 100 μg
For Use With (Application) Blocking, Neutralization
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.