Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human ProBNP Peptide, Synthetic, >95% Purity, 100 μg

Catalog No. 502710931
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0931 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0931 Supplier Abcam Supplier No. AB42238100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Synthetic human proBNP blocking peptide with >95% purity.

  • Protein Name: proBNP
  • Type: Synthetic
  • Purity: >95%
  • Form: Lyophilized; Constituents: 0.1% Acetic acid
  • Tested Applications: BL
  • Species Reactivity: Human
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Peptide-based Substrate
Reactivity Human
Shipping Condition Blue Ice
Target Molecule proBNP (Human)
Label or Dye Unconjugated
Buffer 0.1% acetic acid
Concentration 1 mg/mL
Content And Storage Store at -20°C.
Form Lyophilized
Cross Reactivity Human
Formulation 0.1% Acetic acid
Quantity 100 μg
For Use With (Application) Blocking
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.