Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Procalcitonin ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB016826 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016826 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016826 Supplier R&D Systems™ Supplier No. NBP281267

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. This kit recognizes Human Procalcitonin in samples.

No significant cross-reactivity or interference between Human Procalcitonin and analogues was observed.

Specifications

Assay Range 12.5 to 800 pg/mL
Assay Sensitivity 7.5 pg/mL
Product Line Novus
Product Type ELISA Kit (Chemiluminescence)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 796
Gene Symbol CALCA
Host Species Human
Quantity 1 Kit
Research Discipline Cancer, Cellular Markers, Neuroscience, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Procalcitonin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.