Learn More
Novus Biologicals™ Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Description
Specifications
Specifications
| Assay Range | 15.63 to 1000 pg/mL |
| Assay Sensitivity | 9.38 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | The Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) recognizes Human P1NP in samples. No significant cross-reactivity or interference between Human P1NP and analogues was observed. |
| Gene ID (Entrez) | 1277 |
| Host Species | Human |
| Quantity | 1 Kit |
| Research Discipline | Cellular Markers, Extracellular Matrix, Metastasis |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.