Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Prokineticin-2 Recombinant Protein, PeproTech®

Catalog No. 10046250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10046250UG 250 μg
10046100UG 100 μg
100-46-1MG 1 mg
100-46-20UG 20 μg
10046500UG 500 μg
100-46-50UG 50 μg
100-46-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 10046250UG Supplier Gibco™ Supplier No. 10046250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Prokineticin-2 is an 8.8 kDa protein consisting of 81 amino acid residues, including ten cysteine residues that can potentially form five pairs of intra-molecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Prokineticin-2 (PK2) is a cysteine-rich secreted protein that is expressed in the suprachiasmatic nucleus (SCN) with receptors located in the critical autonomic control centers of the brain. It has a depolarizing effect on neurons expressing the receptor. PK2 is predominantly controlled by the endogenous circadian clock, but light also plays a modulatory role. PK2 functions as a critical SCN output molecule responsible for circadian locomotor rhythms. PK2 expression is high during the day, and responsive to nocturnal light pulses. PK2 also functions as a chemoattractant for subventricular zone-derived neuronal progenitors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HC23
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 60675
Molecular Weight (g/mol) 8.8 kDa
Name Human Prokineticin-2
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Bombina variegata 8 kD protein; Bombina variegata 8 kDa protein; Bv 8 homolog (Bombina variegata); BV8; Bv8/prokineticin 2-like protein; Bv8/prokineticin 2-like protein splice variant; HH4; homolog of mouse Bv8 (Bombina variegata 8 kDa); homolog of mouse Bv8 (Bombina variegata 8 kDa); prokineticin 2; KAL4; MIT1; PK2; Prok1; Prok2; prokineticin 1; prokineticin 2; prokineticin 2 beta; prokineticin-2; Protein Bv8 homolog; secreted protein
Common Name PROK2
Gene Symbol PROK2
Conjugate Unconjugated
Recombinant Recombinant
Sequence AVITGACDKD SQCGGGMCCA VSIWVKSIRI CTPMGKLGDS CHPLTRKVPF FGRRMHHTCP CLPGLACLRT SFNRFICLAQ K
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.