Learn More
Kingfisher Biotech Inc Human Prolactin Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1576H005
Encompass
The Human Prolactin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human Prolactin applications are for cell culture, ELISA standard, and Western Blot Control. Human Prolactin yeast-derived recombinant protein can be purchased in multiple sizes. Human Prolactin Specifications: (Molecular Weight: 22.9 kDa) (Amino Acid Sequence: LPICPGGAAR CQVTLRDLFD RAVVLSHYIH NLSSEMFSEF DKRYTHGRGF ITKAINSCHT SSLATPEDKE QAQQMNQKDF LSLIVSILRS WNEPLYHLVT EVRGMQEAPE AILSKAVEIE EQTKRLLEGM ELIVSQVHPE TKENEIYPVW SGLPSLQMAD EESRLSAYYN LLHCLRRDSH KIDNYLKLLK CRIIHNNNC (199)) (Gene ID: 5617). For research use only. Made in the USA
Catalog No. 50-253-3139
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.