Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Proprotein Convertase 2/PCSK2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6018
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6018 Supplier R&D Systems Supplier No. MAB6018
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proprotein Convertase 2/PCSK2 Monoclonal specifically detects Proprotein Convertase 2/PCSK2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Proprotein Convertase 2/PCSK2
Applications Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 694009
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P16519
Gene Alias EC 3.4.21, EC 3.4.21.94, KEX2-like endoprotease 2, NEC 2, NEC2, NEC2SPC2, neuroendocrine convertase 2, PC2, PC2NEC-2, PCSK2, Prohormone convertase 2, Proprotein convertase 2, proprotein convertase subtilisin/kexin type 2, SPC2
Gene Symbols PCSK2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Proprotein Convertase 2/PCSK2 Gly110-Asn638 Accession # P16519
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5126
Test Specificity Detects human Proprotein Convertase 2/PCSK2 in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Proprotein Convertase 1, 7, 9, or rhFurin is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.