Learn More
R&D Systems™ Human Proprotein Convertase 7 (NP_004707.2) VersaClone cDNA
Shop All Novus Biologicals Products

Specifications
Specifications
| Gene | Proprotein Convertase 7/PCSK7 |
| Accession Number | NP_004707.2 |
| Gene ID (Entrez) | 9159 |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, EC 3.4.21.61, EC 3.4.21.75, EC 3.4.21.93, hPC8, LPC, LPCFLJ23503, Lymphoma proprotein convertase, PC7FLJ45329, PC8prohormone convertase PC7, PCSK7, Prohormone convertase 7, Proprotein convertase 7, Proprotein convertase 8, proprotein convertase subtilisin/kexin type 7, SPC7, SPC7proprotein convertase PC7, Subtilisin/kexin-like protease PC7 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.