Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit
SDP

Catalog No. SPC900 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SPC900 1 Pk. (6 Plates) 6 x 96-well Plates
DPC900 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SPC900 Supplier R&D Systems™ Supplier No. SPC900

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Proprotein Convertase 9/PCSK9 in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.63 to 40 ng/mL
Assay Sensitivity 0.219 ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human PCSK9. This assay recognizes free and LDLR-bound PCSK9.
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 255738
Gene Symbol PCSK9
Quantity 1 Pk. (6 Plates)
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1 convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
Target Proprotein Convertase 9/PCSK9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.