Learn More
Biomatik Corporation Human Protein Phosphatase 2, Regulatory Subunit B'' Alpha (PPP2R3a) ELISA Kit
Supplier:  Biomatik Corporation EKU12029
Encompass_Preferred
Human Protein Phosphatase 2, Regulatory Subunit B'' Alpha (PPP2R3a) ELISA Kit for detecting PPP2R3a in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06190. Gene ID: 5523. Target Synonyms: PPP2R3; PR130; PR72; Serine/Threonine-Protein Phosphatase 2A Regulatory Subunit B Subunit Alpha; Serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5301
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.