Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRPF3 Partial ORF (NP_004689, 585 a.a. - 683 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-334 25 μg
89-958-333 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-334 Supplier Abnova™ Supplier No. H00009129Q01L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. This gene product is one of several proteins that associate with U4 and U6 snRNPs. Mutations in this gene are associated with retinitis pigmentosa-18. [provided by RefSeq]

Sequence: EGGPKAQKKFKRLMLHRIKWDEQTSNTKGDDDEESDEEAVKKTNKCVLVWEGTAKDRSFGEMKFKQCPTENMAREHFKKHGAEHYWDLALSESVLESTD

Specifications

Accession Number NP_004689
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9129
Molecular Weight (g/mol) 36.63kDa
Name PRPF3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen EGGPKAQKKFKRLMLHRIKWDEQTSNTKGDDDEESDEEAVKKTNKCVLVWEGTAKDRSFGEMKFKQCPTENMAREHFKKHGAEHYWDLALSESVLESTD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HPRP3/HPRP3P/PRP3/Prp3p/RP18
Common Name PRPF3
Gene Symbol PRPF3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.