Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PSMB8 Full-length ORF (NP_004150.1, 1 a.a. - 272 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958541 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-541 10 ug
89-958-542 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-541 Supplier Abnova™ Supplier No. H00005696P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. This gene is located in the class II region of the MHC (major histocompatibility complex). Expression of this gene is induced by gamma interferon and this gene product replaces catalytic subunit 3 (proteasome beta 5 subunit) in the immunoproteasome. Proteolytic processing is required to generate a mature subunit. Two alternative transcripts encoding two isoforms have been identified; both isoforms are processed to yield the same mature subunit. [provided by RefSeq]

Sequence: MLIGTPTPRDTTPSSWLTSSLLVEAAPLDDTTLPTPVSSGCPGLEPTEFFQSLGGDGERNVQIEMAHGTTTLAFKFQHGVIAAVDSRASAGSYISALRVNKVIEINPYLLGTMSGCAADCQYWERLLAKECRLYYLRNGERISVSAASKLLSNMMCQYRGMGLSMGSMICGWDKKGPGLYYVDEHGTRLSGNMFSTGSGNTYAYGVMDSGYRPNLSPEEAYDLGRRAIAYATHRDSYSGGVVNMYHMKEDGWVKVESTDVSDLLHQYREANQ

Specifications

Accession Number NP_004150.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5696
Molecular Weight (g/mol) 56.2kDa
Name PSMB8 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MLIGTPTPRDTTPSSWLTSSLLVEAAPLDDTTLPTPVSSGCPGLEPTEFFQSLGGDGERNVQIEMAHGTTTLAFKFQHGVIAAVDSRASAGSYISALRVNKVIEINPYLLGTMSGCAADCQYWERLLAKECRLYYLRNGERISVSAASKLLSNMMCQYRGMGLSMGSMICGWDKKGPGLYYVDEHGTRLSGNMFSTGSGNTYAYGVMDSGYRPNLSPEEAYDLGRRAIAYATHRDSYSGGVVNMYHMKEDGWVKVESTDVSDLLHQYREANQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias D6S216/D6S216E/LMP7/MGC1491/PSMB5i/RING10/beta5i
Common Name PSMB8
Gene Symbol PSMB8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.