Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PTEN Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7250054
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB66551SP 25 μg
MAB66551100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB66551SP Supplier R&D Systems Supplier No. MAB66551SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTEN Monoclonal antibody specifically detects PTEN in Human samples. It is validated for Immunocytochemistry, KnockDown

Specifications

Antigen PTEN
Applications Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 1042838
Conjugate Unconjugated
Dilution Immunocytochemistry 3 -25 ug/mL, Knockout Validated 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P60484
Gene Alias BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM
Host Species Mouse
Immunogen E. coli-derived recombinant human PTEN, Thr2-Val403, Accession # P60484
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5728
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.