Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PTK9 Partial ORF (AAH22344, 80 a.a. - 252 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-751 10 ug
89-958-752 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-751 Supplier Abnova™ Supplier No. H00005756Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1. [provided by RefSeq]

Sequence: VAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSSAKPKGPAGKRGIRRLIRGPAETEATTD

Specifications

Accession Number AAH22344
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5756
Molecular Weight (g/mol) 44.66kDa
Name PTK9 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen VAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSSAKPKGPAGKRGIRRLIRGPAETEATTD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias A6/MGC23788/MGC41876/PTK9
Common Name TWF1
Gene Symbol TWF1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.