Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PTPN12 Full-length ORF (NP_002826.2, 1 a.a. - 780 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958803 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-803 10 ug
89-958-804 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-803 Supplier Abnova™ Supplier No. H00005782P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains a C-terminal PEST motif, which serves as a protein-protein interaction domain, and may regulate protein intracellular half-life. This PTP was found to bind and dephosphorylate the product of the oncogene c-ABL and thus may play a role in oncogenesis. This PTP was also shown to interact with, and dephosphorylate, various products related to cytoskeletal structure and cell adhesion, such as p130 (Cas), CAKbeta/PTK2B, PSTPIP1, and paxillin. This suggests it has a regulatory role in controlling cell shape and mobility. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MEQVEILRKFIQRVQAMKSPDHNGEDNFARDFMRLRRLSTKYRTEKIYPTATGEKEENVKKNRYKDILPFDHSRVKLTLKTPSQDSDYINANFIKGVYGPKAYVATQGPLANTVIDFWRMIWEYNVVIIVMACREFEMGRKKCERYWPLYGEDPITFAPFKISCEDEQARTDYFIRTLLLEFQNESRRLYQFHYVNWPDHDVPSSFDSILDMISLMRKYQEHEDVPICIHCSAGCGRTGAICAIDYTWNLLKAGKIPEEFNVFNLIQEMRTQRHSAVQTKEQYELVHRAIAQLFEKQLQLYEIHGAQKIADGVNEINTENMISSIEPEKQDSPPPKPPRTRSCLVEGDAKEEILQPPEPHPVPPILTPSPPSAFPTVTTVWQDNDRYHPKPVLHMVSSEQHSADLNRNYSKSTELPGKNESTIEQIDKKLERNLSFEIKKVPLQEGPKSFDGNTLLNRGHAIKIKSASPCIADKISKPQELSSDLNVGDTSQNSCVDCSVTQSNKVSVTPPEESQNSDTPPRPDRLPLDEKGHVTWSFHGPENAIPIPDLSEGNSSDINYQTRKTVSLTPSPTTQVETPDLVDHDNTSPLFRTPLSFTNPLHSDDSDSDERNSDGAVTQNKTNISTASATVSAATSTESISTRKVLPMSIARHNIAGTTHSGAEKDVDVSEDSPPPLPERTPESFVLASEHNTPVRSEWSELQSQERSEQKKSEGLITSENEKCDHPAGGIHYEMCIECPPTFSDKREQISENPTEATDIGFGNRCGKPKGPRDPPSEWT

Specifications

Accession Number NP_002826.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5782
Molecular Weight (g/mol) 114.5kDa
Name PTPN12 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MEQVEILRKFIQRVQAMKSPDHNGEDNFARDFMRLRRLSTKYRTEKIYPTATGEKEENVKKNRYKDILPFDHSRVKLTLKTPSQDSDYINANFIKGVYGPKAYVATQGPLANTVIDFWRMIWEYNVVIIVMACREFEMGRKKCERYWPLYGEDPITFAPFKISCEDEQARTDYFIRTLLLEFQNESRRLYQFHYVNWPDHDVPSSFDSILDMISLMRKYQEHEDVPICIHCSAGCGRTGAICAIDYTWNLLKAGKIPEEFNVFNLIQEMRTQRHSAVQTKEQYELVHRAIAQLFEKQLQLYEIHGAQKIADGVNEINTENMISSIEPEKQDSPPPKPPRTRSCLVEGDAKEEILQPPEPHPVPPILTPSPPSAFPTVTTVWQDNDRYHPKPVLHMVSSEQHSADLNRNYSKSTELPGKNESTIEQIDKKLERNLSFEIKKVPLQEGPKSFDGNTLLNRGHAIKIKSASPCIADKISKPQELSSDLNVGDTSQNSCVDCSVTQSNKVSVTPPEESQNSDTPPRPDRLPLDEKGHVTWSFHGPENAIPIPDLSEGNSSDINYQTRKTVSLTPSPTTQVETPDLVDHDNTSPLFRTPLSFTNPLHSDDSDSDERNSDGAVTQNKTNISTASATVSAATSTESISTRKVLPMSIARHNIAGTTHSGAEKDVDVSEDSPPPLPERTPESFVLASEHNTPVRSEWSELQSQERSEQKKSEGLITSENEKCDHPAGGIHYEMCIECPPTFSDKREQISENPTEATDIGFGNRCGKPKGPRDPPSEWT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PTP-PEST/PTPG1
Common Name PTPN12
Gene Symbol PTPN12
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.