Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PTPN13 Partial ORF (NP_006255, 66 a.a. - 175 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958805 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-805 10 ug
89-958-806 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-805 Supplier Abnova™ Supplier No. H00005783Q01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported. [provided by RefSeq]

Sequence: FTDENISNQDLRAFTAPEVLQNQSLTSLSDVEKIHIYSLGMTLYWGADYEVPQSQPIKLGDHLNSILLGMCEDVIYARVSVRTVLDACSAHIRNSNCAPSFSYVKHLVKL

Specifications

Accession Number NP_006255
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5783
Molecular Weight (g/mol) 37.84kDa
Name PTPN13 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen FTDENISNQDLRAFTAPEVLQNQSLTSLSDVEKIHIYSLGMTLYWGADYEVPQSQPIKLGDHLNSILLGMCEDVIYARVSVRTVLDACSAHIRNSNCAPSFSYVKHLVKL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686J1497/FAP-1/PNP1/PTP-BAS/PTP-BL/PTP1E/PTPL1/PTPLE
Common Name PTPN13
Gene Symbol PTPN13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.