Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PTPN4 Full-length ORF (NP_002821.1, 1 a.a. - 926 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-824 10 ug
89-958-825 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-824 Supplier Abnova™ Supplier No. H00005775P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. This PTP has been shown to interact with glutamate receptor delta 2 and epsilon subunits, and is thought to play a role in signalling downstream of the glutamate receptors through tyrosine dephosphorylation. [provided by RefSeq]

Sequence: MTSRFRLPAGRTYNVRASELARDRQHTEVVCNILLLDNTVQAFKVNKHDQGQVLLDVVFKHLDLTEQDYFGLQLADDSTDNPRWLDPNKPIRKQLKRGSPYSLNFRVKFFVSDPNKLQEEYTRYQYFLQIKQDILTGRLPCPSNTAALLASFAVQSELGDYDQSENLSGYLSDYSFIPNQPQDFEKEIAKLHQQHIGLSPAEAEFNYLNTARTLELYGVEFHYARDQSNNEIMIGVMSGGILIYKNRVRMNTFPWLKIVKISFKCKQFFIQLRKELHESRETLLGFNMVNYRACKNLWKACVEHHTFFRLDRPLPPQKNFFAHYFTLGSKFRYCGRTEVQSVQYGKEKANKDRVFARSPSKPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDGKPPALPPKQSKKNSWNQIHYSHSQQDLESHINETFDIPSSPEKPTPNGGIPHDNLVLIRMKPDENGRFGFNVKGGYDQKMPVIVSRVAPGTPADLCVPRLNEGDQVVLINGRDIAEHTHDQVVLFIKASCERHSGELMLLVRPNAVYDVVEEKLENEPDFQYIPEKAPLDSVHQDDHSLRESMIQLAEGLITGTVLTQFDQLYRKKPGMTMSCAKLPQNISKNRYRDISPYDATRVILKGNEDYINANYINMEIPSSSIINQYIACQGPLPHTCTDFWQMTWEQGSSMVVMLTTQVERGRVKCHQYWPEPTGSSSYGCYQVTCHSEEGNTAYIFRKMTLFNQEKNESRPLTQIQYIAWPDHGVPDDSSDFLDFVCHVRNKRAGKEEPVVVHCSAGIGRTGVLITMETAMCLIECNQPVYPLDIVRTMRDQRAMMIQTPSQYRFVCEAILKVYEEGFVKPLTTSTNK

Specifications

Accession Number NP_002821.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5775
Molecular Weight (g/mol) 132.3kDa
Name PTPN4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MTSRFRLPAGRTYNVRASELARDRQHTEVVCNILLLDNTVQAFKVNKHDQGQVLLDVVFKHLDLTEQDYFGLQLADDSTDNPRWLDPNKPIRKQLKRGSPYSLNFRVKFFVSDPNKLQEEYTRYQYFLQIKQDILTGRLPCPSNTAALLASFAVQSELGDYDQSENLSGYLSDYSFIPNQPQDFEKEIAKLHQQHIGLSPAEAEFNYLNTARTLELYGVEFHYARDQSNNEIMIGVMSGGILIYKNRVRMNTFPWLKIVKISFKCKQFFIQLRKELHESRETLLGFNMVNYRACKNLWKACVEHHTFFRLDRPLPPQKNFFAHYFTLGSKFRYCGRTEVQSVQYGKEKANKDRVFARSPSKPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDGKPPALPPKQSKKNSWNQIHYSHSQQDLESHINETFDIPSSPEKPTPNGGIPHDNLVLIRMKPDENGRFGFNVKGGYDQKMPVIVSRVAPGTPADLCVPRLNEGDQVVLINGRDIAEHTHDQVVLFIKASCERHSGELMLLVRPNAVYDVVEEKLENEPDFQYIPEKAPLDSVHQDDHSLRESMIQLAEGLITGTVLTQFDQLYRKKPGMTMSCAKLPQNISKNRYRDISPYDATRVILKGNEDYINANYINMEIPSSSIINQYIACQGPLPHTCTDFWQMTWEQGSSMVVMLTTQVERGRVKCHQYWPEPTGSSSYGCYQVTCHSEEGNTAYIFRKMTLFNQEKNESRPLTQIQYIAWPDHGVPDDSSDFLDFVCHVRNKRAGKEEPVVVHCSAGIGRTGVLITMETAMCLIECNQPVYPLDIVRTMRDQRAMMIQTPSQYRFVCEAILKVYEEGFVKPLTTSTNK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PTPMEG/PTPMEG1
Common Name PTPN4
Gene Symbol PTPN4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.