Learn More
Sino Biological CD45 / PTPRC Antibody (Biotin), Mouse MAb 20ug
Supplier:  Sino Biological 10086MM05B20
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD45 (rh CD45; Catalog#10086-H02H; NP_563578.2; Met1-Lys416). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography, and then biotinylated.
Catalog No. 50-255-5487
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.