Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RAB2B Partial ORF (NP_116235, 127 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959094 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-094 10 μg
89-959-095 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-094 Supplier Abnova™ Supplier No. H00084932Q01S

Used for AP, Array, ELISA, WB-Re

Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rab proteins are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking; see MIM 179508.[supplied by OMIM]

Sequence: RDVKREEGEAFAREHGLIFMETSAKTACNVEEAFINTAKEIYRKIQQGLFDVHNEANGIKIGPQQSISTSVGPSASQRNSRDIGSNSGCC

Specifications

Accession Number NP_116235
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84932
Molecular Weight (g/mol) 35.64kDa
Name RAB2B (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen RDVKREEGEAFAREHGLIFMETSAKTACNVEEAFINTAKEIYRKIQQGLFDVHNEANGIKIGPQQSISTSVGPSASQRNSRDIGSNSGCC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ14824
Common Name RAB2B
Gene Symbol RAB2B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.