Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RAD17 Partial ORF (AAH32304, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959226 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-226 10 ug
89-959-227 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-226 Supplier Abnova™ Supplier No. H00005884Q01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is highly similar to the gene product of Schizosaccharomyces pombe rad17, a cell cycle checkpoint gene required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein shares strong similarity with DNA replication factor C (RFC), and can form a complex with RFCs. This protein binds to chromatin prior to DNA damage and is phosphorylated by the checkpoint kinase ATR following damage. This protein recruits the RAD1-RAD9-HUS1 checkpoint protein complex onto chromatin after DNA damage, which may be required for its phosphorylation. The phosphorylation of this protein is required for the DNA-damage-induced cell cycle G2 arrest, and is thought to be a critical early event during checkpoint signaling in DNA-damaged cells. Eight alternatively spliced transcript variants of this gene, which encode four distinct proteins, have been reported. Two pseudogenes, located on chromosomes 7 and 13, have been identified. [provided by RefSeq]

Sequence: MNQVTDWVDPSFDDFLECSGVSTITATSLGVNNSSHRRKNGPSTLESSRFPARKRGNLSSLEQIYGLENSKEYLSENEPWVDKYKPETQHELAVHKKKIEEVETWLKAQV

Specifications

Accession Number AAH32304
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5884
Molecular Weight (g/mol) 37.73kDa
Name RAD17 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MNQVTDWVDPSFDDFLECSGVSTITATSLGVNNSSHRRKNGPSTLESSRFPARKRGNLSSLEQIYGLENSKEYLSENEPWVDKYKPETQHELAVHKKKIEEVETWLKAQV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CCYC/FLJ41520/HRAD17/R24L/RAD17SP/RAD24
Common Name RAD17
Gene Symbol RAD17
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.