Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RAPGEF5 Partial ORF (AAH39203, 345 a.a. - 444 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-375 25 μg
89-959-374 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-375 Supplier Abnova™ Supplier No. H00009771Q01L

Used for AP, Array, ELISA, WB-Re

Members of the RAS (see HRAS; MIM 190020) subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RAPGEF5, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation (Rebhun et al., 2000 [PubMed 10934204]).[supplied by OMIM]

Sequence: AYRDAFKKMKPPKIPFVPLLLKDVTFIHEGNKTFLDNLVNFEKLHMIADTVRTLRHCRTNQFGDLSPKEHQELKSYVNHLYVIDSQQALFELSHRIEPRV

Specifications

Accession Number AAH39203
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9771
Molecular Weight (g/mol) 36.63kDa
Name RAPGEF5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen AYRDAFKKMKPPKIPFVPLLLKDVTFIHEGNKTFLDNLVNFEKLHMIADTVRTLRHCRTNQFGDLSPKEHQELKSYVNHLYVIDSQQALFELSHRIEPRV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GFR/KIAA0277/MR-GEF/REPAC
Common Name RAPGEF5
Gene Symbol RAPGEF5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.