Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RARRES1 Full-length ORF (NP_002879.2, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959400 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-400 10 ug
89-959-401 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-400 Supplier Abnova™ Supplier No. H00005918P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWVRKT

Specifications

Accession Number NP_002879.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5918
Molecular Weight (g/mol) 50.71kDa
Name RARRES1 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 10 ug
Immunogen MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWVRKT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias TIG1
Common Name RARRES1
Gene Symbol RARRES1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.