Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RARRES3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6139
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6139 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6139 Supplier R&D Systems Supplier No. MAB6139
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RARRES3 Monoclonal specifically detects RARRES3 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen RARRES3
Applications Immunocytochemistry
Classification Monoclonal
Clone 659209
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UL19
Gene Alias HRASLS4, PLA1/2-3, RAR-responsive protein TIG3, retinoic acid receptor responder (tazarotene induced) 3, retinoic acid receptor responder protein 3, retinoic acid-inducible gene 1, Retinoid-inducible gene 1 protein, RIG1, Tazarotene-induced gene 3 protein, TIG3, TIG3MGC8906
Gene Symbols RARRES3
Host Species Mouse
Immunogen E. coli-derived recombinant human RARRES3 Asp12-Lys134 Accession # Q9UL19
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5920
Test Specificity Recognizes human RARRES3 in Western blots and direct ELISAs. In Western blots and direct ELISAs, no cross-reactivity with recombinant human (rh) RARRES1 or rhLatexin is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.