Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RASAL1 Full-length ORF (AAH14420.1, 1 a.a. - 776 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959427 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-427 10 μg
89-959-428 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-427 Supplier Abnova™ Supplier No. H00008437P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. These proteins stimulate the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This particular family member contains domains which are characteristic of the GAP1 subfamily of RasGAP proteins but, in contrast to the other GAP1 family members, this protein is strongly and selectively expressed in endocrine tissues. [provided by RefSeq]

Sequence: MAKSSSLNVRVVEGRALPAKDVSGSSDPYCLVKVDDEVVARTATVWRSLGPFWGEEYTVHLPLDFHQLAFYVLDEDTVGHDDIIGKISLSREAITADPRGIDSWINLSRVDPDAEVQGEICLSVQMLEDGQGRCLRCHVLQARDLAPRDISGTSDPFARVFWGSQSLETSTIKKTRFPHWDEVLELREMPGAPSPLRVELWDWDMVGKNDFLGMVEFSPKTLQQKPPKGWFRLLPFPRAEEDSGGNLGALRVKVRLIEDRVLPSQCYQPLMELLMESVQGPAEEDTASPLALLEELTLGDCRQDLATKLVKLFLGRGLAGHFLDYLTRREVARTMDPNTLFRSNSLASKSMEQFMKLVGMPYLHEVLKPVISRVFEEKKYMELDPCKMDLGRTRRISFKGALSEEQMRETSLGLLTGYLGPIVDAIVGSVGRCPPAMRLAFKQLHRRVEERFPQAEHQDVKYLAISGFLFLRFFAPAILTPKLFDLRDQHADPQTSRSLLLLAKAVQSIGNLGQQLGQGKELWMAPLHPFLLQCVSRVRDFLDRLVDVDGDEAGVPARALFPPSAIVREGYLLKRKEEPAGLATRFAFKKRYVWLSGETLSFSKSPEWQVVTQDGTGALHTTYLQCKNVNELNQWLSALRKASAPNPNKLAACHPGAFRSARWTCCLQAERSAAGCSRTHSAVTLGDWSDPLDPDAEAQTVYRQLLLGRDQLRLKLLEDSNMDTTLEADTGACPEVLARQRAATARLLEVLADLDRAHEEFQQQERGKAALGPLGP

Specifications

Accession Number AAH14420.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8437
Molecular Weight (g/mol) 113.2kDa
Name RASAL1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAKSSSLNVRVVEGRALPAKDVSGSSDPYCLVKVDDEVVARTATVWRSLGPFWGEEYTVHLPLDFHQLAFYVLDEDTVGHDDIIGKISLSREAITADPRGIDSWINLSRVDPDAEVQGEICLSVQMLEDGQGRCLRCHVLQARDLAPRDISGTSDPFARVFWGSQSLETSTIKKTRFPHWDEVLELREMPGAPSPLRVELWDWDMVGKNDFLGMVEFSPKTLQQKPPKGWFRLLPFPRAEEDSGGNLGALRVKVRLIEDRVLPSQCYQPLMELLMESVQGPAEEDTASPLALLEELTLGDCRQDLATKLVKLFLGRGLAGHFLDYLTRREVARTMDPNTLFRSNSLASKSMEQFMKLVGMPYLHEVLKPVISRVFEEKKYMELDPCKMDLGRTRRISFKGALSEEQMRETSLGLLTGYLGPIVDAIVGSVGRCPPAMRLAFKQLHRRVEERFPQAEHQDVKYLAISGFLFLRFFAPAILTPKLFDLRDQHADPQTSRSLLLLAKAVQSIGNLGQQLGQGKELWMAPLHPFLLQCVSRVRDFLDRLVDVDGDEAGVPARALFPPSAIVREGYLLKRKEEPAGLATRFAFKKRYVWLSGETLSFSKSPEWQVVTQDGTGALHTTYLQCKNVNELNQWLSALRKASAPNPNKLAACHPGAFRSARWTCCLQAERSAAGCSRTHSAVTLGDWSDPLDPDAEAQTVYRQLLLGRDQLRLKLLEDSNMDTTLEADTGACPEVLARQRAATARLLEVLADLDRAHEEFQQQERGKAALGPLGP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias RASAL
Common Name RASAL1
Gene Symbol RASAL1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.