Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human/Rat Neuritin Recombinant Protein, PeproTech®

Catalog No. 45036D1MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-36D-1MG 1 mg
45036D100UG 100 μg
45036D20UG 20 μg
45036D250UG 250 μg
45036D500UG 500 μg
450-36D-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 450-36D-1MG Supplier Gibco™ Supplier No. 45036D1MG
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human/Rat Neuritin is a covalently disulfide-linked homodimer, consisting of two 9.72 kDa polypeptide monomers, each containing 88 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Neuritin belongs to the neuritin family and is expressed in postmitotic-differentiating neurons of the developmental nervous system as well as neuronal structures associated with plasticity in adults. Its expression can be induced by neural activity and neurotrophins. The protein contains a consensus cleavage signal found in glycosylphoshatidylinositol (GPI)-anchored proteins and promotes neurite outgrowth and arborization, indicating its role in promoting neuritogenesis. Overexpression of the protein may be associated with astrocytoma progression. Alternative splicing leads to multiple transcript variants. NRN1 is associated with Severe Congenital Neutropenia 8 and Spinal Muscular Atrophy, Type III.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O08957, Q9NPD7
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 51299, 83834
Molecular Weight (g/mol) 19.4 kDa
Name Human/Rat Neuritin
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 0710008J23Rik; Candidate plasticity gene 15 protein; candidate plasticity-related gene 15; Cpg15; dJ380B8.2; Neuritin; neuritin 1; NRN; NRN1
Common Name Neuritin
Gene Symbol NRN1
Biological Activity Determined by a cell proliferation using rat C6 cells. The ED50 for this effect is 20.0-30.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence AGKCDAVFKG FSDCLLKLGD SMANYPQGLD DKTNIKTVCT YWEDFHSCTV TALTDCQEGA KDMWDKLRKE SKNLNIQGSL FELCGSGN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.