Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat P-Selectin/CD62P Alexa Fluor Plus 488-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10138259
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10138259 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10138259 Supplier R&D Systems Supplier No. AF137AFP488100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

P-Selectin/CD62P Polyclonal antibody specifically detects P-Selectin/CD62P in Human,Rat samples. It is validated for Western Blot,Adhesion Blockade

Specifications

Antigen P-Selectin/CD62P
Applications Western Blot, Adhesion Blockade
Classification Polyclonal
Conjugate Alexa Fluor Plus 488
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP
Host Species Sheep
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6403
Target Species Human, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Product Type Antibody
Form Liquid
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.