Learn More
Human/Rat SUSD2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | SUSD2 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1279D |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide |
| Gene Accession No. | Q9UGT4 |
| Gene Alias | BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.