Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Rat Total HIF-2 alpha/EPAS1 DuoSet IC ELISA

Catalog No. DYC29972
Change view
Click to view available options
Packaging Quantity:
2 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC29972 2 Plates
DYC29975 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DYC29972 Supplier R&D Systems™ Supplier No. DYC29972
This item is not returnable. View return policy

Human/Rat Total HIF-2 alpha/EPAS1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human/Rat Total HIF-2 alpha/EPAS1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, rat HIF-2 alpha/EPAS1.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human, Rat
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 2034
Synonym Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF 2A, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF2 alpha, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.