Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RBBP8 Full-length ORF (NP_002885.1, 1 a.a. - 897 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959503 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-503 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-959-503 Supplier Abnova™ Supplier No. H00005932P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a ubiquitously expressed nuclear protein. It is found among several proteins that bind directly to retinoblastoma protein, which regulates cell proliferation. This protein complexes with transcriptional co-repressor CTBP. It is also associated with BRCA1 and is thought to modulate the functions of BRCA1 in transcriptional regulation, DNA repair, and/or cell cycle checkpoint control. It is suggested that this gene may itself be a tumor suppressor acting in the same pathway as BRCA1. Three transcript variants encoding two different isoforms have been found for this gene. More transcript variants exist, but their full-length natures have not been determined. [provided by RefSeq]

Sequence: MNISGSSCGSPNSADTSSDFKDLWTKLKECHDREVQGLQVKVTKLKQERILDAQRLEEFFTKNQQLREQQKVLHETIKVLEDRLRAGLCDRCAVTEEHMRKKQQEFENIRQQNLKLITELMNERNTLQEENKKLSEQLQQKIENDQQHQAAELECEEDVIPDSPITAFSFSGVNRLRRKENPHVRYIEQTHTKLEHSVCANEMRKVSKSSTHPQHNPNENEILVADTYDQSQSPMAKAHGTSSYTPDKSSFNLATVVAETLGLGVQEESETQGPMSPLGDELYHCLEGNHKKQPFEESTRNTEDSLRFSDSTSKTPPQEELPTRVSSPVFGATSSIKSGLDLNTSLSPSLLQPGKKKHLKTLPFSNTCISRLEKTRSKSEDSALFTHHSLGSEVNKIIIQSSNKQILINKNISESLGEQNRTEYGKDSNTDKHLEPLKSLGGRTSKRKKTEEESEHEVSCPQASFDKENAFPFPMDNQFSMNGDCVMDKPLDLSDRFSAIQRQEKSQGSETSKNKFRQVTLYEALKTIPKGFSSSRKASDGNCTLPKDSPGEPCSQECIILQPLNKCSPDNKPSLQIKEENAVFKIPLRPRESLETENVLDDIKSAGSHEPIKIQTRSDHGGCELASVLQLNPCRTGKIKSLQNNQDVSFENIQWSIDPGADLSQYKMDVTVIDTKDGSQSKLGGETVDMDCTLVSETVLLKMKKQEQKGEKSSNEERKMNDSLEDMFDRTTHEEYESCLADSFSQAADEEEELSTATKKLHTHGDKQDKVKQKAFVEPYFKGDERETSLQNFPHIEVVRKKEERRKLLGHTCKECEIYYADMPAEEREKKLASCSRHRFRYIPPNTPENFWEVGFPSTQTCMERGYIKEDLDPCPRPKRRQPYNAIFSPKGKEQKT

Specifications

Accession Number NP_002885.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5932
Molecular Weight (g/mol) 128.3kDa
Name RBBP8 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MNISGSSCGSPNSADTSSDFKDLWTKLKECHDREVQGLQVKVTKLKQERILDAQRLEEFFTKNQQLREQQKVLHETIKVLEDRLRAGLCDRCAVTEEHMRKKQQEFENIRQQNLKLITELMNERNTLQEENKKLSEQLQQKIENDQQHQAAELECEEDVIPDSPITAFSFSGVNRLRRKENPHVRYIEQTHTKLEHSVCANEMRKVSKSSTHPQHNPNENEILVADTYDQSQSPMAKAHGTSSYTPDKSSFNLATVVAETLGLGVQEESETQGPMSPLGDELYHCLEGNHKKQPFEESTRNTEDSLRFSDSTSKTPPQEELPTRVSSPVFGATSSIKSGLDLNTSLSPSLLQPGKKKHLKTLPFSNTCISRLEKTRSKSEDSALFTHHSLGSEVNKIIIQSSNKQILINKNISESLGEQNRTEYGKDSNTDKHLEPLKSLGGRTSKRKKTEEESEHEVSCPQASFDKENAFPFPMDNQFSMNGDCVMDKPLDLSDRFSAIQRQEKSQGSETSKNKFRQVTLYEALKTIPKGFSSSRKASDGNCTLPKDSPGEPCSQECIILQPLNKCSPDNKPSLQIKEENAVFKIPLRPRESLETENVLDDIKSAGSHEPIKIQTRSDHGGCELASVLQLNPCRTGKIKSLQNNQDVSFENIQWSIDPGADLSQYKMDVTVIDTKDGSQSKLGGETVDMDCTLVSETVLLKMKKQEQKGEKSSNEERKMNDSLEDMFDRTTHEEYESCLADSFSQAADEEEELSTATKKLHTHGDKQDKVKQKAFVEPYFKGDERETSLQNFPHIEVVRKKEERRKLLGHTCKECEIYYADMPAEEREKKLASCSRHRFRYIPPNTPENFWEVGFPSTQTCMERGYIKEDLDPCPRPKRRQPYNAIFSPKGKEQKT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CTIP/RIM
Common Name RBBP8
Gene Symbol RBBP8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.