Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RBM12 Full-length ORF (NP_006038.2, 1 a.a. - 932 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959523 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-523 25 μg
89-959-522 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-523 Supplier Abnova™ Supplier No. H00010137P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5' UTR results in two transcript variants. Both variants encode the same protein. [provided by RefSeq]

Sequence: MAVVIRLQGLPIVAGTMDIRHFFSGLTIPDGGVHIVGGELGEAFIVFATDEDARLGMMRTGGTIKGSKVTLLLSSKTEMQNMIELSRRRFETANLDIPPANASRSGPPPSSGMSSRVNLPTTVSNFNNPSPSVVTATTSVHESNKNIQTFSTASVGTAPPNMGASFGSPTFSSTVPSTASPMNTVPPPPIPPIPAMPSLPPMPSIPPIPVPPPVPTLPPVPPVPPIPPVPSVPPMTPLPPMSGMPPLNPPPVAPLPAGMNGSGAPMNLNNNLNPMFLGPLNPVNPIQMNSQSSVKPLPINPDDLYVSVHGMPFSAMENDVRDFFHGLRVDAVHLLKDHVGRNNGNGLVKFLSPQDTFEALKRNRMLMIQRYVEVSPATERQWVAAGGHITFKQNMGPSGQTHPPPQTLPRSKSPSGQKRSRSRSPHEAGFCVYLKGLPFEAENKHVIDFFKKLDIVEDSIYIAYGPNGKATGEGFVEFRNEADYKAALCRHKQYMGNRFIQVHPITKKGMLEKIDMIRKRLQNFSYDQREMILNPEGDVNSAKVCAHITNIPFSITKMDVLQFLEGIPVDENAVHVLVDNNGQGLGQALVQFKNEDDARKSERLHRKKLNGREAFVHVVTLEDMREIEKNPPAQGKKGLKMPVPGNPAVPGMPNAGLPGVGLPSAGLPGAGLPSTGLPGSAITSAGLPGAGMPSAGIPSAGGEEHAFLTVGSKEANNGPPFNFPGNFGGSNAFGPPIPPPGLGGGAFGDARPGMPSVGNSGLPGLGLDVPGFGGGPNNLSGPSGFGGGPQNFGNGPGSLGGPPGFGSGPPGLGSAPGHLGGPPAFGPGPGPGPGPGPIHIGGPPGFASSSGKPGPTVIKVQNMPFTVSIDEILDFFYGYQVIPGSVCLKYNEKGMPTGEAMVAFESRDEATAAVIDLNDRPIGSRKVKLVLG

Specifications

Accession Number NP_006038.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10137
Molecular Weight (g/mol) 123.8kDa
Name RBM12 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MAVVIRLQGLPIVAGTMDIRHFFSGLTIPDGGVHIVGGELGEAFIVFATDEDARLGMMRTGGTIKGSKVTLLLSSKTEMQNMIELSRRRFETANLDIPPANASRSGPPPSSGMSSRVNLPTTVSNFNNPSPSVVTATTSVHESNKNIQTFSTASVGTAPPNMGASFGSPTFSSTVPSTASPMNTVPPPPIPPIPAMPSLPPMPSIPPIPVPPPVPTLPPVPPVPPIPPVPSVPPMTPLPPMSGMPPLNPPPVAPLPAGMNGSGAPMNLNNNLNPMFLGPLNPVNPIQMNSQSSVKPLPINPDDLYVSVHGMPFSAMENDVRDFFHGLRVDAVHLLKDHVGRNNGNGLVKFLSPQDTFEALKRNRMLMIQRYVEVSPATERQWVAAGGHITFKQNMGPSGQTHPPPQTLPRSKSPSGQKRSRSRSPHEAGFCVYLKGLPFEAENKHVIDFFKKLDIVEDSIYIAYGPNGKATGEGFVEFRNEADYKAALCRHKQYMGNRFIQVHPITKKGMLEKIDMIRKRLQNFSYDQREMILNPEGDVNSAKVCAHITNIPFSITKMDVLQFLEGIPVDENAVHVLVDNNGQGLGQALVQFKNEDDARKSERLHRKKLNGREAFVHVVTLEDMREIEKNPPAQGKKGLKMPVPGNPAVPGMPNAGLPGVGLPSAGLPGAGLPSTGLPGSAITSAGLPGAGMPSAGIPSAGGEEHAFLTVGSKEANNGPPFNFPGNFGGSNAFGPPIPPPGLGGGAFGDARPGMPSVGNSGLPGLGLDVPGFGGGPNNLSGPSGFGGGPQNFGNGPGSLGGPPGFGSGPPGLGSAPGHLGGPPAFGPGPGPGPGPGPIHIGGPPGFASSSGKPGPTVIKVQNMPFTVSIDEILDFFYGYQVIPGSVCLKYNEKGMPTGEAMVAFESRDEATAAVIDLNDRPIGSRKVKLVLG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HRIHFB2091/KIAA0765/SWAN
Common Name RBM12
Gene Symbol RBM12
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.