Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RBP4/Retinol-Binding Protein 4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB33781100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB33781100 100 μg
MAB33781SP 25 μg
MAB33781500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB33781100 Supplier R&D Systems Supplier No. MAB33781100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications ELISA
Classification Monoclonal
Clone 393006
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P02753
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma
Gene Symbols RBP4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human RBP4/Retinol-Binding Protein 4 Glu19-Leu201 Accession # P02753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5950
Test Specificity Detects human RBP4/Retinol-Binding Protein 4 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse RBP4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.