Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RCBTB1 Partial ORF (NP_060661.3, 2 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959634 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-634 25 μg
89-959-633 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-959-634 Supplier Abnova™ Supplier No. H00055213Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein with an N-terminal RCC1 domain and a C-terminal BTB (broad complex, tramtrack and bric-a-brac) domain. In rat, over-expression of this gene in vascular smooth muscle cells induced cellular hypertrophy. In rat, the C-terminus of RCBTB1 interacts with the angiotensin II receptor-1A. In humans, this gene maps to a region of chromosome 13q that is frequently deleted in B-cell chronic lymphocytic leukemia and other lymphoid malignancies. [provided by RefSeq]

Sequence: VDVGKWPIFTLLSPQEIASIRKACVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLS

Specifications

Accession Number NP_060661.3
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55213
Molecular Weight (g/mol) 35.53kDa
Name RCBTB1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen VDVGKWPIFTLLSPQEIASIRKACVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CLLD7/CLLL7/GLP/MGC33184/RP11-185C18.1
Common Name RCBTB1
Gene Symbol RCBTB1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.