Learn More
Creative Biomart Laronidase, Recombinant Human IDUA
Supplier:  Creative Biomart THP0132
Encompass_Preferred
Human recombinant alpha-L-iduronidase, 628 residues (mature form), produced by recombinant DNAtechnology in a Chinese hamster ovary cell line. The product is a glycoprotein with a molecular weight of approximately 83 kD. The predicted amino acid sequence of the recombinant form, as well as the nucleotide sequence that encodes it, are identical to a polymorphic form of human a-L-iduronidase. It contains 6 N-linked oligosaccharide modification sites.
Catalog No. 50-208-4183
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.