Learn More
STEMCELL Technologies Human Recombinant FGF-7 (KGF), Size: 10 µg
SDP
Supplier:  STEMCELL Technologies 78046
Encompass_Preferred
Fibroblast growth factor 7 (FGF-7) is a member of the FGF family, and acts exclusively through a subset of FGF receptor isoforms expressed predominantly by epithelial cells (Finch & Rubin). FGF-7 seems to act specifically on epithelial cells and stimulates proliferation, migration, and differentiation of these cells, and also participates in epithelial protection and repair both in vitro and in vivo (Finch & Rubin; Werner). In contrast, FGF-7 is produced solely by cells of mesenchymal origin, and functions as a paracrine mediator of mesenchymal-epithelial communication (Rubin et al.). FGF-7 has also been shown to supplement several wound-healing properties of bioengineered skin (Erdag et al.) and to induce autophagy in human keratinocytes (Belleudi et al.). Additionally, FGF-7 has a role in pluripotent stem cell differentiation to endodermal pancreatic-like insulin-producing cells and thymic epithelial cells (Inami et al.; Niu et al.).
Catalog No. 50-197-6344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.