Learn More
STEMCELL Technologies Human Recombinant Follistatin, Size: 20 µg
SDP
Supplier:  STEMCELL Technologies 78109
Encompass
Follistatin, a glycosylated monomeric protein, is a modulator of transforming growth factor beta (TGF-β) superfamily signaling. It binds to and inhibits the function of activin, myostatin, growth differentiation factors, and bone morphogenetic proteins (BMP) (Hansen & Plomgaard). Follistatin inhibits mesoderm induction, suppresses synthesis and secretion of pituitary follicle-stimulating hormone, regulates liver regeneration, and causes infertility (Guo et al.; Iemura et al.). Follistatin exhibits anti-inflammatory effects, and it could be used as a biomarker in cancer (Hansen & Plomgaard).
Catalog No. 50-197-6487
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.