Learn More
STEMCELL Technologies Human Recombinant GDNF, Size: 10 µg
SDP
Supplier:  STEMCELL Technologies 78058
Encompass_Preferred
Use glial cell line-derived neurotrophic factor (GDNF) to promote the survival and morphological differentiation of midbrain dopaminergic neurons (Granholm et al. J Neurosci, 2000). GDNF is a neurotrophic factor and a member of the tumor growth factor (TGF)-beta superfamily. The GDNF family ligands signal through binding to specific GDNF-family receptor-α co-receptors, activating the RET receptor tyrosine kinase (Durbec et al. Nature, 1996). GDNF, together with Human Recombinant BDNF (Catalog #78005), BrainPhys™ Neuronal Medium (Catalog #05790), and other supplements, can be used to differentiate human pluripotent stem cell-derived neural progenitor cells into neurons (Bardy et al. Proc Natl Acad Sci USA, 2015). Human recombinant GDNF from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 3 μg/mL; endotoxin levels verified ≤ 1 EU/μg protein. Alternative names: Astrocyte-derived trophic factor, ATF, ATF1, ATF2, Glial cell-derived neurotrophic factor.
Catalog No. NC1563608
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.