Learn More
STEMCELL Technologies Human Recombinant IFN-gamma, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 78020.1
Encompass_Preferred
Activate the JAK/STAT signal transduction pathway and control a diverse array of immunological functions using interferon-gamma (IFN-γ) (de Weerd & Nguyen. Immunol Cell Biol, 2012). IFN-γ stimulates the antimicrobial and anti-tumor activity of macrophages, NK cells, and neutrophils by promoting the production of reactive oxygen species, nitric oxide, and complement (Schroder et al. J Leukoc Biol, 2004). IFN-γ also enhances MHC class I and II expression in dendritic cells and mononuclear phagocytes. In B cells, IFN-γ stimulates survival and growth in both mouse and human cells and redirects B cells from proliferation towards differentiation. IFN-γ favors the development of Th1 vs Th2 cells and stimulates monocyte differentiation and function (Schroder et al.). Human recombinant IFN-γ from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 0.05 ng/mL; endotoxin levels verified ≤ 0.2 EU/μg protein. Alternative names: Interferon gamma, Type II interferon.
Catalog No. 50-197-6285
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.