Learn More
STEMCELL Technologies Human Recombinant LIF, Size: 10 µg
SDP
Supplier:  STEMCELL Technologies 78055.1
Encompass_Preferred
Use leukemia inhibitory factor (LIF) to regulate a variety of developmental functions. LIF is an interleukin 6 class cytokine that regulates proliferation, invasion, and differentiation of trophoblasts following blastocyst attachment (Suman et al. JAK-STAT, 2014). Upon binding of LIF to LIF receptor (LIFR), LIFR associates with gp130 and activates JAK/STAT and MAPK signaling (Suman et al.). LIFR activation of STAT3 is essential for maintaining the mouse embryonic stem cell phenotype (Niwa et al. Genes Dev, 1998). Human LIF can be used for the maintenance of mouse embryonic stem cells (Dahéron et al. Stem Cells, 2004). Human recombinant LIF from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 0.2 ng/mL; endotoxin levels verified ≤ 0.2 EU/μg protein. Alternative names: CDF, Cholinergic differentiation factor, D Factor, DIA, Differentiation-inducing factor, Differentiation inhibitory activity, Emfilermin, Hepatocyte-stimulating factor III, HILDA, Leukemia inhibitory factor, MLPLI.
Catalog No. 50-197-6365
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.